4,6-Dibrom-2,3-dichloranilin

Lieferant: Thermo Scientific

CD07896.1 CD07896.10 CD07896.50
MAYBCD07896.1EA 0 EUR
MAYBCD07896.1 MAYBCD07896.10 MAYBCD07896.50
4,6-Dibrom-2,3-dichloranilin
4,6-Dibrom-2,3-dichloranilin
Formel: C₆H₃Br₂Cl₂N
Molecular Weight: 319.81 g/mol
MDL: MFCD00052815
CAS-Nummer: 113571-15-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR