ω-Brom-4-nitroacetophenon
Lieferant: Thermo Scientific
Synonyme:
2-Brom-4'-nitroacetophenon, 4-Nitrophenacylbromid, Brommethyl-p-nitrophenylketon, α-Brom-4-nitroacetophenon
DP00146.1
DP00146.10
DP00146.50
MAYBDP00146.1EA
0
EUR
MAYBDP00146.1
MAYBDP00146.10
MAYBDP00146.50
ω-Brom-4-nitroacetophenon
ω-Brom-4-nitroacetophenon
Formel:
C₈H₆BrNO₃ Schmelzpunkt: 100 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007356 CAS-Nummer: 99-81-0 EINECS: 202-789-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...