ω-Brom-4-chloracetophenon

Lieferant: CHEMPUR

Synonyme: 4-Chlorphenacylbromid, Brommethyl-p-chlorphenylketon, 2-Brom-4'-chloracetophenon, α-Brom-4-chloracetophenon

DP01340.250 DP01340.1
CHMPDP01340.250EA 0 EUR
CHMPDP01340.250 MAYBDP01340.1
ω-Brom-4-chloracetophenon
ω-Brom-4-chloracetophenon
Formel: C₈H₆BrClO
Molecular Weight: 233,49 g/mol
Siedepunkt: 278 °C (1013 hPa)
Schmelzpunkt: 110 °C
MDL: MFCD00000625
CAS-Nummer: 536-38-9
EINECS: 208-631-5
UN: 3261
ADR: 8,III
Merck Index: 13,02172

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR