ω-Brom-2-chloracetophenon
Lieferant: Thermo Scientific
Synonyme:
2-Brom-2'-chloracetophenon, 2-Chlorphenacylbromid, Brommethyl-o-chlorphenylketon, α-Brom-2-chloracetophenon
SPB06335DA
SPB06335EA
MAYBSPB06335DAEA
0
EUR
MAYBSPB06335DA
MAYBSPB06335EA
ω-Brom-2-chloracetophenon
ω-Brom-2-chloracetophenon
Formel:
C₈H₆BrClO Molecular Weight: 233,49 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00832993 CAS-Nummer: 5000-66-8 UN: 1760 ADR: 8,II |
Learn more
![](/m-de.vwr.com/de_DE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...