4'-Trifluormethoxyacetophenon

Lieferant: Thermo Scientific

Synonyme: 1-[4-(Trifluormethoxy)phenyl]-1-ethanon

TL00401.1 TL00401.10 TL00401.50
MAYBTL00401.1EA 0 EUR
MAYBTL00401.1 MAYBTL00401.10 MAYBTL00401.50
4'-Trifluormethoxyacetophenon
4'-Trifluormethoxyacetophenon
Formel: C₉H₇F₃O₂
Molecular Weight: 204,15 g/mol
Siedepunkt: 110 °C (30 mmHg)
Dichte: 1,27 g/cm³ (20 °C)
MDL: MFCD00042404
CAS-Nummer: 85013-98-5
EINECS: 285-066-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR