2-Brom-4-methyl-6-nitroanilin

Lieferant: COMBI-BLOCKS

Synonyme: 2-Brom-6-nitro-p-toluidin, 4-Amino-3-brom-5-nitrotoluol

AN-1051-5G AN-1051-25G AN-1051-1G
COBBAN-1051-5GEA 50.9 EUR
COBBAN-1051-5G COBBAN-1051-25G COBBAN-1051-1G
2-Brom-4-methyl-6-nitroanilin
2-Brom-4-methyl-6-nitroanilin
Formel: C₇H₇BrN₂O₂
MDL: MFCD00209452
CAS-Nummer: 827-24-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR