2-Brom-5-fluor-4-methylanilin

Lieferant: COMBI-BLOCKS

Synonyme: 2-Brom-5-fluor-p-toluidin

HC-2066-5G HC-2066-25G HC-2066-1G
COBBHC-2066-5GEA 323 EUR
COBBHC-2066-5G COBBHC-2066-25G COBBHC-2066-1G
2-Brom-5-fluor-4-methylanilin
2-Brom-5-fluor-4-methylanilin
Formel: C₇H₇BrFN
Molecular Weight: 204.04 g/mol
MDL: MFCD00142871
CAS-Nummer: 202865-78-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR