3-Brom-5-(trifluormethyl)anilin

Lieferant: COMBI-BLOCKS
Achtung

HC-6321-5G HC-6321-25G HC-6321-100G
COBBHC-6321-5GEA 0 EUR
COBBHC-6321-5G COBBHC-6321-25G COBBHC-6321-100G
3-Brom-5-(trifluormethyl)anilin
3-Brom-5-(trifluormethyl)anilin
Formel: C₇H₅BrF₃N
MDL: MFCD00236205
CAS-Nummer: 54962-75-3
EINECS: 259-412-6
UN: 2810
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR