Propargylamin
Lieferant: COMBI-BLOCKS
Synonyme:
3-Aminopropin
OS-7456-100G
OS-7456-25G
COBBOS-7456-100GEA
460
EUR
COBBOS-7456-100G
COBBOS-7456-25G
Propargylamin
Propargylamin
Formel:
C₃H₅N Molecular Weight: 55,08 g/mol Siedepunkt: 83 °C (1013 hPa) Dichte: 0,867 g/cm³ (16 °C) |
MDL:
MFCD00008198 CAS-Nummer: 2450-71-7 EINECS: 219-513-8 UN: 3286 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...