3-Brom-4-chloranilin

Lieferant: COMBI-BLOCKS

OS-7488-25G OS-7488-5G OS-7488-1G
COBBOS-7488-25GEA 235 EUR
COBBOS-7488-25G COBBOS-7488-5G COBBOS-7488-1G
3-Brom-4-chloranilin
3-Brom-4-chloranilin
Formel: C₆H₅BrClN
Molecular Weight: 206.47 g/mol
MDL: MFCD00173905
CAS-Nummer: 823-54-1
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR