Kalium(brommethyl)trifluoroborat

Lieferant: COMBI-BLOCKS

Synonyme: (Brommethyl)trifluorborsäure Kaliumsalz

TB-0018-25G TB-0018-1G TB-0018-5G
COBBTB-0018-25GEA 307 EUR
COBBTB-0018-25G COBBTB-0018-1G COBBTB-0018-5G
Kalium(brommethyl)trifluoroborat
Kalium(brommethyl)trifluoroborat
Formel: CH₂BBrF₃K
Molecular Weight: 200.83 g/mol
MDL: MFCD09265154
CAS-Nummer: 888711-44-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR