Kaliummethyltrifluoroborat
Lieferant: COMBI-BLOCKS
Synonyme:
Methyltrifluorborsäure Kaliumsalz
TB-0028-1G
TB-0028-25G
TB-0028-5G
COBBTB-0028-1GEA
33.2
EUR
COBBTB-0028-1G
COBBTB-0028-25G
COBBTB-0028-5G
Kaliummethyltrifluoroborat
Kaliummethyltrifluoroborat
Formel:
CH₃BF₃K Molecular Weight: 121,94 g/mol |
MDL:
MFCD08276757 CAS-Nummer: 13862-28-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...