Kalium(4-chlor-3-nitrophenyl)trifluoroborat

Lieferant: COMBI-BLOCKS

TB-2037-5G TB-2037-1G
COBBTB-2037-5GEA 692 EUR
COBBTB-2037-5G COBBTB-2037-1G
Kalium(4-chlor-3-nitrophenyl)trifluoroborat
Kalium(4-chlor-3-nitrophenyl)trifluoroborat
Formel: C₆H₃BClF₃KNO₂
Molecular Weight: 263.45 g/mol
MDL: MFCD12026115
CAS-Nummer: 1218908-71-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR