Kalium(4-cyanphenyl)trifluoroborat

Lieferant: COMBI-BLOCKS

Synonyme: (4-Cyanophenyl)trifluoroboric acid potassium salt

TB-3201-1G TB-3201-25G TB-3201-5G
COBBTB-3201-1GEA 88.9 EUR
COBBTB-3201-1G COBBTB-3201-25G COBBTB-3201-5G
Kalium(4-cyanphenyl)trifluoroborat
Kalium(4-cyanphenyl)trifluoroborat
Formel: C₇H₄BF₃KN
Molecular Weight: 209.02 g/mol
MDL: MFCD04115736
CAS-Nummer: 850623-36-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR