Kalium(3-nitrophenyl)trifluoroborat

Lieferant: COMBI-BLOCKS

TB-3205-5G TB-3205-1G TB-3205-25G
COBBTB-3205-5GEA 0 EUR
COBBTB-3205-5G COBBTB-3205-1G COBBTB-3205-25G
Kalium(3-nitrophenyl)trifluoroborat
Kalium(3-nitrophenyl)trifluoroborat
Formel: C₆H₄BF₃KNO₂
MDL: MFCD03701627
CAS-Nummer: 192863-40-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR