Kalium(3-methylphenyl)trifluoroborat
Lieferant: COMBI-BLOCKS
Synonyme:
Kalium(m-tolyl)trifluoroborat, (3-Methylphenyl)trifluorborsäure Kaliumsalz
TB-3206-5G
TB-3206-25G
TB-3206-1G
COBBTB-3206-5GEA
149
EUR
COBBTB-3206-5G
COBBTB-3206-25G
COBBTB-3206-1G
Kalium(3-methylphenyl)trifluoroborat
Kalium(3-methylphenyl)trifluoroborat
Formel:
C₇H₇BF₃K Molecular Weight: 198.04 g/mol |
MDL:
MFCD04115739 CAS-Nummer: 850623-42-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...