Kalium(1-naphthyl)trifluoroborat

Lieferant: COMBI-BLOCKS

Synonyme: (1-Naphthyl)trifluorborsäure Kaliumsalz

TB-3216-1G TB-3216-25G TB-3216-5G
COBBTB-3216-1GEA 88.6 EUR
COBBTB-3216-1G COBBTB-3216-25G COBBTB-3216-5G
Kalium(1-naphthyl)trifluoroborat
Kalium(1-naphthyl)trifluoroborat
Formel: C₁₀H₇BF₃K
Molecular Weight: 234.07 g/mol
MDL: MFCD06659936
CAS-Nummer: 166328-07-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR