Kalium(3-chlor-4-fluorphenyl)trifluoroborat

Lieferant: COMBI-BLOCKS

Synonyme: (3-Chlor-4-fluorphenyl)trifluorborsäure Kaliumsalz

TB-3222-1G TB-3222-5G TB-3222-25G
COBBTB-3222-1GEA 66 EUR
COBBTB-3222-1G COBBTB-3222-5G COBBTB-3222-25G
Kalium(3-chlor-4-fluorphenyl)trifluoroborat
Kalium(3-chlor-4-fluorphenyl)trifluoroborat
Formel: C₆H₃BClF₄K
Molecular Weight: 236,44 g/mol
MDL: MFCD04115752
CAS-Nummer: 850623-59-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR