Kalium(4-ethoxyphenyl)trifluoroborat

Lieferant: COMBI-BLOCKS

Synonyme: (4-Ethoxyphenyl)trifluorborsäure Kaliumsalz

TB-3223-1G TB-3223-5G
COBBTB-3223-1GEA 91.2 EUR
COBBTB-3223-1G COBBTB-3223-5G
Kalium(4-ethoxyphenyl)trifluoroborat
Kalium(4-ethoxyphenyl)trifluoroborat
Formel: C₈H₉BF₃KO
Molecular Weight: 228.06 g/mol
MDL: MFCD04115753
CAS-Nummer: 850623-60-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR