Kalium(3-chlorphenyl)trifluoroborat
Lieferant: COMBI-BLOCKS
Synonyme:
(3-Chlorphenyl)trifluorborsäure Kaliumsalz
TB-3224-1G
TB-3224-5G
COBBTB-3224-1GEA
63
EUR
COBBTB-3224-1G
COBBTB-3224-5G
Kalium(3-chlorphenyl)trifluoroborat
Kalium(3-chlorphenyl)trifluoroborat
Formel:
C₆H₄BClF₃K Molecular Weight: 218.45 g/mol |
MDL:
MFCD04115754 CAS-Nummer: 411206-75-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...