Kalium(2-methoxyphenyl)trifluoroborat
Lieferant: COMBI-BLOCKS
Synonyme:
(2-Methoxyphenyl)trifluorborsäure Kaliumsalz
TB-3227-1G
TB-3227-25G
TB-3227-5G
COBBTB-3227-1GEA
49.7
EUR
COBBTB-3227-1G
COBBTB-3227-25G
COBBTB-3227-5G
Kalium(2-methoxyphenyl)trifluoroborat
Kalium(2-methoxyphenyl)trifluoroborat
Formel:
C₇H₇BF₃KO Molecular Weight: 214,04 g/mol |
MDL:
MFCD04115756 CAS-Nummer: 236388-46-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...