Kalium(2-methylphenyl)trifluoroborat
Lieferant: COMBI-BLOCKS
Synonyme:
(2-Methylphenyl)trifluorborsäure Kaliumsalz, Kalium(o-tolyl)trifluoroborat
TB-3228-1G
TB-3228-25G
TB-3228-5G
COBBTB-3228-1GEA
74.2
EUR
COBBTB-3228-1G
COBBTB-3228-25G
COBBTB-3228-5G
Kalium(2-methylphenyl)trifluoroborat
Kalium(2-methylphenyl)trifluoroborat
Formel:
C₇H₇BF₃K Molecular Weight: 198,04 g/mol |
MDL:
MFCD03790006 CAS-Nummer: 274257-34-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...