Kalium(4-methoxyphenyl)trifluoroborat
Lieferant: COMBI-BLOCKS
Synonyme:
(4-Methoxyphenyl)trifluorborsäure Kaliumsalz
TB-3231-1G
TB-3231-25G
TB-3231-5G
COBBTB-3231-1GEA
51.3
EUR
COBBTB-3231-1G
COBBTB-3231-25G
COBBTB-3231-5G
Kalium(4-methoxyphenyl)trifluoroborat
Kalium(4-methoxyphenyl)trifluoroborat
Formel:
C₇H₇BF₃KO Molecular Weight: 214,04 g/mol |
MDL:
MFCD04112717 CAS-Nummer: 192863-36-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...