Kalium(4-methylphenyl)trifluoroborat

Lieferant: COMBI-BLOCKS

Synonyme: Kalium(p-tolyl)trifluoroborat, (4-Methylphenyl)trifluorborsäure Kaliumsalz

TB-3234-5G TB-3234-25G TB-3234-1G
COBBTB-3234-5GEA 141 EUR
COBBTB-3234-5G COBBTB-3234-25G COBBTB-3234-1G
Kalium(4-methylphenyl)trifluoroborat
Kalium(4-methylphenyl)trifluoroborat
Formel: C₇H₇BF₃K
Molecular Weight: 198,04 g/mol
MDL: MFCD01318171
CAS-Nummer: 216434-82-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR