Kalium(3,4-dichlorphenyl)trifluoroborat
Lieferant: COMBI-BLOCKS
Synonyme:
(3,4-Dichlorphenyl)trifluorborsäure Kaliumsalz
TB-3238-1G
TB-3238-25G
TB-3238-5G
COBBTB-3238-1GEA
73.9
EUR
COBBTB-3238-1G
COBBTB-3238-25G
COBBTB-3238-5G
Kalium(3,4-dichlorphenyl)trifluoroborat
Kalium(3,4-dichlorphenyl)trifluoroborat
Formel:
C₆H₃BCl₂F₃K Molecular Weight: 252,9 g/mol |
MDL:
MFCD04115763 CAS-Nummer: 850623-68-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...