Kalium(4-nitrophenyl)trifluoroborat

Lieferant: COMBI-BLOCKS

TB-3241-1G TB-3241-5G
COBBTB-3241-1GEA 0 EUR
COBBTB-3241-1G COBBTB-3241-5G
Kalium(4-nitrophenyl)trifluoroborat
Kalium(4-nitrophenyl)trifluoroborat
Formel: C₆H₄BF₃KNO₂
MDL: MFCD04115766
CAS-Nummer: 850623-71-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR