Kalium(4-chlorphenyl)trifluoroborat

Lieferant: COMBI-BLOCKS

Synonyme: (4-Chlorphenyl)trifluorborsäure Kaliumsalz

TB-3246-1G TB-3246-25G TB-3246-5G
COBBTB-3246-1GEA 58.4 EUR
COBBTB-3246-1G COBBTB-3246-25G COBBTB-3246-5G
Kalium(4-chlorphenyl)trifluoroborat
Kalium(4-chlorphenyl)trifluoroborat
Formel: C₆H₄BClF₃K
Molecular Weight: 218.45 g/mol
MDL: MFCD04115770
CAS-Nummer: 661465-44-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR