Kalium(4-fluorphenyl)trifluoroborat

Lieferant: COMBI-BLOCKS

TB-3248-1G TB-3248-25G TB-3248-5G
COBBTB-3248-1GEA 0 EUR
COBBTB-3248-1G COBBTB-3248-25G COBBTB-3248-5G
Kalium(4-fluorphenyl)trifluoroborat
Kalium(4-fluorphenyl)trifluoroborat
Formel: C₆H₄BF₄K
MDL: MFCD01318170
CAS-Nummer: 192863-35-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR