Kalium(2-fluorphenyl)trifluoroborat

Lieferant: COMBI-BLOCKS

Synonyme: (2-Fluorphenyl)trifluorborsäure Kaliumsalz

TB-3251-5G TB-3251-1G TB-3251-25G
COBBTB-3251-5GEA 303 EUR
COBBTB-3251-5G COBBTB-3251-1G COBBTB-3251-25G
Kalium(2-fluorphenyl)trifluoroborat
Kalium(2-fluorphenyl)trifluoroborat
Formel: C₆H₄BF₄K
Molecular Weight: 202 g/mol
MDL: MFCD06659937
CAS-Nummer: 166328-10-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR