Kalium(3-fluorphenyl)trifluoroborat
Lieferant: COMBI-BLOCKS
Synonyme:
(3-Fluorphenyl)trifluorborsäure Kaliumsalz
TB-3252-5G
TB-3252-1G
TB-3252-25G
COBBTB-3252-5GEA
136
EUR
COBBTB-3252-5G
COBBTB-3252-1G
COBBTB-3252-25G
Kalium(3-fluorphenyl)trifluoroborat
Kalium(3-fluorphenyl)trifluoroborat
Formel:
C₆H₄BF₄K Molecular Weight: 202 g/mol |
MDL:
MFCD06659938 CAS-Nummer: 267006-24-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...