Kalium(3,5-difluorphenyl)trifluoroborat
Lieferant: COMBI-BLOCKS
Synonyme:
(3,5-Difluorphenyl)trifluorborsäure Kaliumsalz
TB-3253-1G
TB-3253-25G
TB-3253-5G
COBBTB-3253-1GEA
88.6
EUR
COBBTB-3253-1G
COBBTB-3253-25G
COBBTB-3253-5G
Kalium(3,5-difluorphenyl)trifluoroborat
Kalium(3,5-difluorphenyl)trifluoroborat
Formel:
C₆H₃BF₅K Molecular Weight: 219,99 g/mol |
MDL:
MFCD06659939 CAS-Nummer: 267006-26-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...