Kalium(4-bromphenyl)trifluoroborat

Lieferant: COMBI-BLOCKS

Synonyme: (4-Bromphenyl)trifluorborsäure Kaliumsalz

TB-3254-1G TB-3254-25G TB-3254-5G
COBBTB-3254-1GEA 41 EUR
COBBTB-3254-1G COBBTB-3254-25G COBBTB-3254-5G
Kalium(4-bromphenyl)trifluoroborat
Kalium(4-bromphenyl)trifluoroborat
Formel: C₆H₄BBrF₃K
Molecular Weight: 262,91 g/mol
MDL: MFCD02093978
CAS-Nummer: 374564-35-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR