1H,1H-Pentadecafluor-1-octanol

Lieferant: FLUOROCHEM

001240-5G 001240-25G
FLUO001240-5GEA 48.8 EUR
FLUO001240-5G FLUO001240-25G
1H,1H-Pentadecafluor-1-octanol
1H,1H-Pentadecafluor-1-octanol
Formel: C₈H₃F₁₅O
MDL: MFCD00004675
CAS-Nummer: 307-30-2
EINECS: 206-197-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR