1-Ethoxy-2,2,2-trifluorethanol
Lieferant: FLUOROCHEM
Synonyme:
Trifluoracetaldehyd-Ethylhalbacetal, Trifluoracetaldehydethylhemiacetal
001243-25G
001243-100G
FLUO001243-25GEA
107
EUR
FLUO001243-25G
FLUO001243-100G
1-Ethoxy-2,2,2-trifluorethanol
1-Ethoxy-2,2,2-trifluoroethanol
Formel:
C₄H₇F₃O₂ Molecular Weight: 144.09 g/mol |
MDL:
MFCD00000441 CAS-Nummer: 433-27-2 EINECS: 207-087-6 UN: 1989 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...