1-Methoxy-2,2,2-trifluorethanol

Lieferant: FLUOROCHEM

Synonyme: Trifluoracetaldehyd-Methylhalbacetal, Trifluoracetaldehydmethylhemiacetal

001527-25G 001527-100G 001527-10G
FLUO001527-25GEA 83.1 EUR
FLUO001527-25G FLUO001527-100G FLUO001527-10G
1-Methoxy-2,2,2-trifluorethanol
1-Methoxy-2,2,2-trifluoroethanol
Formel: C₃H₅F₃O₂
Molecular Weight: 130,07 g/mol
MDL: MFCD00013572
CAS-Nummer: 431-46-9
EINECS: 207-072-4
UN: 3271
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR