1-Brom-4,4,4-trifluorbutan

Lieferant: FLUOROCHEM

001531.5G 001531-1G
FLUO001531.5GEA 224 EUR
FLUO001531.5G FLUO001531-1G
1-Brom-4,4,4-trifluorbutan
1-Bromo-4,4,4-trifluorobutane
Formel: C₄H₆BrF₃
Molecular Weight: 190,99 g/mol
MDL: MFCD00066335
CAS-Nummer: 406-81-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR