3'-Trifluormethoxyacetophenon
Lieferant: FLUOROCHEM
Synonyme:
1-[3-(Trifluormethoxy)phenyl]-1-ethanon
002123-1G
002123-5G
002123-25G
FLUO002123-1GEA
55.4
EUR
FLUO002123-1G
FLUO002123-5G
FLUO002123-25G
3'-Trifluormethoxyacetophenon
3'-Trifluormethoxyacetophenon
Formel:
C₉H₇F₃O₂ Molecular Weight: 204.15 |
MDL:
MFCD00042403 CAS-Nummer: 170141-63-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...