1H,1H-Tricosafluor-1-dodecanol

Lieferant: FLUOROCHEM

002298-5G 002298-25G
FLUO002298-5GEA 118 EUR
FLUO002298-5G FLUO002298-25G
1H,1H-Tricosafluor-1-dodecanol
1H,1H-Tricosafluor-1-dodecanol
Formel: C₁₂H₃F₂₃O
MDL: MFCD00153235
CAS-Nummer: 423-65-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR