1H,1H-Heptacosafluor-1-tetradecanol

Lieferant: FLUOROCHEM

002299-5G 002299-25G
FLUO002299-5GEA 101 EUR
FLUO002299-5G FLUO002299-25G
1H,1H-Heptacosafluor-1-tetradecanol
1H,1H-Heptacosafluor-1-tetradecanol
Formel: C₁₄H₃F₂₇O
MDL: MFCD00153264
CAS-Nummer: 15622-57-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR