1H,1H,10H,10H-Hexadecafluor-1,10-decandiol

Lieferant: FLUOROCHEM

Synonyme: 1H,1H,10H,10H-Perfluor-1,10-decandiol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-Hexadecafluor-1,10-decandiol

002307-5G 002307-25G
FLUO002307-5GEA 84.9 EUR
FLUO002307-5G FLUO002307-25G
1H,1H,10H,10H-Hexadecafluor-1,10-decandiol
1H,1H,10H,10H-Hexadecafluor-1,10-decandiol
Formel: C₁₀H₆F₁₆O₂
Molecular Weight: 462,13 g/mol
MDL: MFCD00192200
CAS-Nummer: 754-96-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR