1,4-Dibromoctafluorbutan
Lieferant: FLUOROCHEM
002313-5G
002313-25G
002313-100G
FLUO002313-5GEA
137
EUR
FLUO002313-5G
FLUO002313-25G
FLUO002313-100G
1,4-Dibromoctafluorbutan
1,4-Dibromooctafluorobutane
Formel:
C₄Br₂F₈ |
MDL:
MFCD00153112 CAS-Nummer: 335-48-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...