1H,1H,7H-Dodecafluor-1-heptanol

Lieferant: FLUOROCHEM

Synonyme: 1H,1H,7H-Perfluor-1-heptanol, 2,2,3,3,4,4,5,5,6,6,7,7-Dodecafluor-1-heptanol

003212-25G 003212-100G 003212-500G
FLUO003212-25GEA 31.1 EUR
FLUO003212-25G FLUO003212-100G FLUO003212-500G
1H,1H,7H-Dodecafluor-1-heptanol
1H,1H,7H-Dodecafluor-1-heptanol
Formel: C₇H₄F₁₂O
Molecular Weight: 332,09 g/mol
MDL: MFCD00039630
CAS-Nummer: 335-99-9
EINECS: 206-406-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR