1H,1H,11H-Eicosafluor-1-unundecanol
Lieferant: FLUOROCHEM
Synonyme:
2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11-Eicosafluor-1-undecanol, 1H,1H,11H-Perfluor-1-undecanol
003214-5G
003214-25G
FLUO003214-5GEA
0
EUR
FLUO003214-5G
FLUO003214-25G
1H,1H,11H-Eicosafluor-1-unundecanol
1H,1H,11H-Eicosafluor-1-unundecanol
Formel:
C₁₁H₄F₂₀O Molecular Weight: 532,12 g/mol |
MDL:
MFCD00039628 CAS-Nummer: 307-70-0 EINECS: 206-209-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...