2-Chlor-1,4-dibrom-1,1,2-trifluorbutan

Lieferant: FLUOROCHEM

Synonyme: 1,4-Dibromo-2-chloro-1,1,2-trifluorobutane

004388-1G 004388-5G
FLUO004388-1GEA 31.1 EUR
FLUO004388-1G FLUO004388-5G
2-Chlor-1,4-dibrom-1,1,2-trifluorbutan
2-Chloro-1,4-dibromo-1,1,2-trifluorobutane
Formel: C₄H₄Br₂ClF₃
Molecular Weight: 304,33 g/mol
MDL: MFCD00017985
CAS-Nummer: 378-13-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR