ω-Brom-3,4-difluoracetophenon
Lieferant: FLUOROCHEM
Synonyme:
2-Brom-3',4'-difluoracetophenon, α-Brom-3,4-difluoracetophenon, 3,4-Difluorphenacylbromid, 2-Brom-1-(3,4-difluorphenyl)ethan-1-on
004848-5G
004848-1G
004848-25G
FLUO004848-5GEA
0
EUR
FLUO004848-5G
FLUO004848-1G
FLUO004848-25G
ω-Brom-3,4-difluoracetophenon
ω-Brom-3,4-difluoracetophenon
Formel:
C₈H₅BrF₂O |
MDL:
MFCD00085010 CAS-Nummer: 40706-98-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...