1H,1H,2H,2H-Tridecafluor-1-octanol

Lieferant: FLUOROCHEM

Synonyme: 1H,1H,2H,2H-Perfluor-1-octanol, 3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluor-1-octanol

007128-25G 007128-100G
FLUO007128-25GEA 97.2 EUR
FLUO007128-25G FLUO007128-100G
1H,1H,2H,2H-Tridecafluor-1-octanol
1H,1H,2H,2H-Tridecafluor-1-octanol
Formel: C₈H₅F₁₃O
Molecular Weight: 364,11 g/mol
MDL: MFCD00042143
CAS-Nummer: 647-42-7
EINECS: 211-477-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR