2-Phenyl-1,1,1-trifluor-3-butin-2-ol
Lieferant: FLUOROCHEM
Synonyme:
α-Ethinyl-α-trifluormethylbenzylalkohol, 1,1,1-Trifluor-2-phenyl-3-butin-2-ol
008692-1G
008692-5G
FLUO008692-1GEA
0
EUR
FLUO008692-1G
FLUO008692-5G
2-Phenyl-1,1,1-trifluor-3-butin-2-ol
2-Phenyl-1,1,1-trifluor-3-butin-2-ol
Formel:
C₁₀H₇F₃O Molecular Weight: 200.16 g/mol |
MDL:
MFCD00792447 CAS-Nummer: 99727-20-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...