4,4,5,5,6,6,6-Heptafluor-1-hexanol
Lieferant: FLUOROCHEM
Synonyme:
4,4,5,5,6,6,6-Heptafluorohexan-1-ol
010475-25G
010475-5G
FLUO010475-25GEA
0
EUR
FLUO010475-25G
FLUO010475-5G
4,4,5,5,6,6,6-Heptafluor-1-hexanol
4,4,5,5,6,6,6-Heptafluoro-1-hexanol
Formel:
C₆H₇F₇O Molecular Weight: 228.11 g/mol |
MDL:
MFCD00077554 CAS-Nummer: 679-02-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...