2-Brom-5-fluor-4-methylanilin
Lieferant: CHEMPUR
Synonyme:
2-Brom-5-fluor-p-toluidin
FL13241.POR
013241-1G
013241-5G
CHMPFL13241.POREA
0
EUR
CHMPFL13241.POR
FLUO013241-1G
FLUO013241-5G
2-Brom-5-fluor-4-methylanilin
2-Brom-5-fluor-4-methylanilin
Formel:
C₇H₇BrFN Molecular Weight: 204.04 g/mol |
MDL:
MFCD00142871 CAS-Nummer: 202865-78-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...