3'-Chlor-4'-methoxyacetophenon
Lieferant: FLUOROCHEM
Synonyme:
1-(3-Chlor-4-methoxyphenyl)ethan-1-on, 1-(3-Chlor-4-methoxyphenyl)ethanon
013883-25G
013883-5G
FLUO013883-25GEA
1430
EUR
FLUO013883-25G
FLUO013883-5G
3'-Chlor-4'-methoxyacetophenon
3'-Chlor-4'-methoxyacetophenon
Formel:
C₉H₉ClO₂ Molecular Weight: 184,62 g/mol |
MDL:
MFCD04973413 CAS-Nummer: 37612-52-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...